Fmoc-N-Me-Ile-OH 98%

Supplier: Ambeed
Warning

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-isoleucine, Fmoc-N-Me-Ile-OH, N-Fmoc-N-methyl-L-isoleucine

A294599-25G A294599-100G A294599-10G A294599-250MG A294599-1G
103814-016EA 6 USD
103814-016 103835-156 76837-792 77002-380 77002-382 77002-384 77305-658
Fmoc-N-Me-Ile-OH 98%
Fmoc-N-Me-Ile-OH

Precaución: for Research Use Only

Formula: C₂₂H₂₅NO₄
MW: 367.445 g/mol
Boiling Pt: 537.3 °C (760 mm Hg)
Melting Pt: 177…183 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153389
CAS Number: 138775-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR