Fmoc-N-Me-Glu(OtBu)-OH 95%

Supplier: Ambeed
Warning

A229043-10G A229043-1G A229043-25G A229043-5G
103819-444EA 35 USD
103819-444 103850-344 77000-144 77089-164 77089-166 77089-168
Fmoc-N-Me-Glu(OtBu)-OH 95%
Fmoc-N-Me-Glu(OtBu)-OH

Precaución: for Research Use Only

Formula: C₂₅H₂₉NO₆
Boiling Pt: 612.3 °C (760 mm Hg)
Melting Pt: 116…119 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00237028
CAS Number: 200616-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR