Fmoc-N-Me-D-Ala-OH 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-N-methyl-D-alanine

A123882-100G A123882-100MG A123882-10G A123882-250MG A123882-25G A123882-1G A123882-5G
103800-458EA 11 USD
103800-458 76797-556 76797-558 76797-560 76797-562 76797-564 77312-580 77312-582
Fmoc-N-Me-D-Ala-OH 97%
Fmoc-N-Me-D-Ala-OH

Precaución: for Research Use Only

Formula: C₁₉H₁₉NO₄
MW: 325.364 g/mol
Boiling Pt: 514.9 °C (760 mm Hg)
Flash Pt: 265.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153385
CAS Number: 138774-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR