N-Fmoc-L-norleucine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Nle-OH, FMOC-L-Norleucine

A200227-100G A200227-10G A200227-5G A200227-1G A200227-25G A200227-500G
103817-176EA 6 USD
103817-176 103847-804 76873-362 76873-364 77085-864 77302-666 77302-668 77671-772
N-Fmoc-L-norleucine 97%
N-Fmoc-L-norleucine

Precaución: for Research Use Only

Formula: C₂₁H₂₃NO₄
MW: 353.4 g/mol
Boiling Pt: 565.6 °C (760 mm Hg)
Melting Pt: 141…144 °C
Flash Pt: 295.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00037537
CAS Number: 77284-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR