Fmoc-N-amido-PEG10-Acid 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-NH-PEG10-CH2CH2COOH

A100154-100MG A100154-10G A100154-250MG A100154-5G
103795-272EA 124 USD
103795-272 103820-988 77245-060 77245-062 77245-064 77282-444
Fmoc-N-amido-PEG10-Acid 97%
Fmoc-N-amido-PEG10-Acid

Precaución: for Research Use Only

Formula: C₃₈H₅₇NO₁₄
Storage Temperature: Refrigerator
MDL Number: MFCD28122954
CAS Number: 2101563-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR