Fmoc-N-(2,4,6-trimethoxybenzyl)-glycine 97%

Supplier: Ambeed
Danger

A821972-100MG A821972-10G A821972-250MG A821972-1G A821972-25G A821972-5G
103841-138EA 70 USD
103841-138 77159-356 77159-358 77159-360 77308-394 77308-396 77308-398
Fmoc-N-(2,4,6-trimethoxybenzyl)-glycine 97%
Fmoc-N-(2,4,6-trimethoxybenzyl)-glycine

Precaución: for Research Use Only

Formula: C₂₇H₂₇NO₇
MW: 477.51 g/mol
Boiling Pt: 676.8 °C (760 mm Hg)
Melting Pt: 148…155 °C
Flash Pt: 363.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09952622
CAS Number: 166881-43-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR