Fmoc-Met(O)-OH 95%

Supplier: Ambeed
Warning

A161334-100G A161334-10G A161334-1G A161334-5G
103805-476EA 14 USD
103805-476 103843-714 77110-706 77110-708 77319-682 77319-684
Fmoc-Met(O)-OH 95%
Fmoc-Met(O)-OH

Precaución: for Research Use Only

Formula: C₂₀H₂₁NO₅S
Storage Temperature: Refrigerator
MDL Number: MFCD00077064
CAS Number: 76265-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR