Fmoc-Lys(Z)-OH 98%

Supplier: Ambeed
Warning

Synonyms: Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-carbobenzoxy-L-lysine, Fmoc-Lys(Cbz)-OH, Fmoc-Lys(Z)-OH, Nalpha-Fmoc-Nepsilon-benzyloxycarbonyl-L-lysine, Nalpha-Fmoc-Nepsilon-Cbz-L-lysine

A113954-100G A113954-10G A113954-25G A113954-1G A113954-250MG
103798-324EA 6 USD
103798-324 103824-166 77271-108 77271-110 77271-112 77310-988 77747-827
Fmoc-Lys(Z)-OH 98%
Fmoc-Lys(Z)-OH

Precaución: for Research Use Only

Formula: C₂₉H₃₀N₂O₆
MW: 502.567 g/mol
Boiling Pt: 751.2 °C (760 mm Hg)
Melting Pt: 113…114 °C
Flash Pt: 408.1 °C
Storage Temperature: Freezer
MDL Number: MFCD00065662
CAS Number: 86060-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR