Fmoc-Lys(Mtt)-OH 98%

Supplier: Ambeed
Warning

A119825-250MG A119825-25G A119825-1G A119825-5G A119825-100G A119825-10G
103799-846EA 12 USD
103799-846 76995-744 76995-746 77297-720 77297-722 77549-406 77549-408
Fmoc-Lys(Mtt)-OH 98%
Fmoc-Lys(Mtt)-OH

Precaución: for Research Use Only

Formula: C₄₁H₄₀N₂O₄
Boiling Pt: 798.78 °C (760 mm Hg)
Flash Pt: 436.887 °C
Storage Temperature: Freezer
MDL Number: MFCD00237166
CAS Number: 167393-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR