N-ɑ-Fmoc-N-ε-boc-L-lysine 98%

Supplier: Ambeed
Warning

A168331-25G A168331-100G A168331-10G A168331-500G A168331-5G A168331-1000G
103844-534EA 7 USD
103844-534 77081-594 77273-610 77273-612 77273-614 77300-926 77567-468
N-ɑ-Fmoc-N-ε-boc-L-lysine 98%
N-alpha-Fmoc-N-epsilon-boc-L-lysine

Precaución: For research use only.

Formula: C₂₆H₃₂N₂O₆
Boiling Pt: 685.7 °C (760 mm Hg)
Melting Pt: 130…135 °C
Flash Pt: 368.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00037138
CAS Number: 71989-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR