Fmoc-Lys(biotin)-OH 97%

Supplier: Ambeed
Warning

Synonyms: Nα-Biotinyl-Nepsilon-Fmoc-L-lysine, Fmoc-Biocytin, (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-6-(5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)hexanoic acid, Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-biotinyl-L-lysine, Fmoc-Lys(biotinyl)-OH, Nalpha-Fmoc-Nepsilon-biotinyl-L-lysine

A178274-250MG A178274-1G A178274-5G A178274-25G A178274-100MG
103807-308EA 73 USD
103807-308 103845-648 76799-608 77083-054 77083-056 77600-442 77667-468
Fmoc-Lys(biotin)-OH 97%
Fmoc-Lys(biotin)-OH

Precaución: for Research Use Only

Formula: C₃₁H₃₈N₄O₆S
Storage Temperature: Refrigerator
MDL Number: MFCD00270741
CAS Number: 146987-10-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR