Fmoc-Lys(Dde)-OH 97%

Supplier: Ambeed
Warning

A183613-1G A183613-5G A183613-10G A183613-250MG A183613-100G
103815-650EA 9 USD
103815-650 103846-212 77083-818 77083-820 77096-942 77096-944 77685-429
Fmoc-Lys(Dde)-OH 97%
Fmoc-Lys(Dde)-OH

Precaución: for Research Use Only

Formula: C₃₁H₃₆N₂O₆
Melting Pt: 80 °C
Storage Temperature: Freezer
MDL Number: MFCD00278818
CAS Number: 150629-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR