N-Fmoc-L-leucine 98%

Supplier: Ambeed
Warning

A132224-100G A132224-500G A132224-5G A132224-1000G A132224-25G A132224-10G
103808-442EA 313 USD
103808-442 77272-054 77272-056 77298-548 77314-272 77314-274 77592-520
N-Fmoc-L-leucine 98%
N-Fmoc-L-leucine

Precaución: for Research Use Only

Formula: C₂₁H₂₃NO₄
MW: 353.4 g/mol
Boiling Pt: 559.8 °C (760 mm Hg)
Melting Pt: 152…156 °C
Flash Pt: 292.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00037133
CAS Number: 35661-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR