Fmoc-Cyclopropylglycine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-Amino(cyclopropyl)acetic acid, N-FMOC protected, Fmoc-L-Cyclopropylglycine
A192277-100MG
A192277-250MG
A192277-1G
103816-432EA
151
USD
103816-432
103847-038
76813-946
76813-948
77084-910
Fmoc-Cyclopropylglycine 95%
Fmoc-Cyclopropylglycine
Precaución: for Research Use Only
Formula:
C₂₀H₁₉NO₄ MW: 337.4 g/mol Boiling Pt: 577.1±33.0 °C (760 mm Hg) Flash Pt: 302.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06659145 CAS Number: 1212257-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...