Fmoc-Inp-OH 98%

Supplier: Ambeed
Warning

Synonyms: 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-piperidinecarboxylic acid, 1-Fmoc-isonipecotic acid, 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]isonipecotic acid, Fmoc-isonipecotic acid, 1-Fmoc-4-piperidinecarboxylic acid, Fmoc-Isn-OH, 1-Fmoc-piperidine-4-carboxylic acid

A122361-100G A122361-10G A122361-1G A122361-25G
103800-210EA 6 USD
103800-210 77009-476 77009-478 77312-344 77611-048
Fmoc-Inp-OH 98%
Fmoc-Inp-OH

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₄
MW: 351.402 g/mol
Boiling Pt: 561.6 °C (760 mm Hg)
Melting Pt: 190 °C (760 mm Hg)
Flash Pt: 293.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00273475
CAS Number: 148928-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR