N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine 98%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-L-isoleucine, Fmoc-Ile-OH, FMOC-L-Isoleucine

A333977-1000G A333977-100G A333977-10G A333977-500G A333977-25G A333977-5G
103810-154EA 379 USD
103810-154 76839-628 77277-860 77277-862 77277-864 77306-462 77593-970
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine 98%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine

Precaución: for Research Use Only

Formula: C₂₁H₂₃NO₄
MW: 353.42 g/mol
Melting Pt: 145…147 °C
Flash Pt: 292.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00037125
CAS Number: 71989-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR