trans-N-Fmoc-4-tert-butoxy-L-proline 97% (by HPLC)

Supplier: Ambeed
Warning

A265743-1G A265743-5G A265743-10G A265743-25G
103812-752EA 6 USD
103812-752 103852-648 77092-352 77092-354 77289-346 77304-932
trans-N-Fmoc-4-tert-butoxy-L-proline 97% (by HPLC)
trans-N-Fmoc-4-tert-butoxy-L-proline

Precaución: for Research Use Only

Formula: C₂₄H₂₇NO₅
Boiling Pt: 578.6 °C (760 mm Hg)
Melting Pt: 63 °C
Flash Pt: 303.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00151930
CAS Number: 122996-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR