(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A139951-250MG
A139951-500G
A139951-25G
A139951-1G
103802-800EA
11
USD
103802-800
76798-192
76798-194
77299-102
77315-908
(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid 98%
(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid
Precaución: for Research Use Only
Formula:
C₂₁H₂₁NO₄ Boiling Pt: 561.6 °C (760 mm Hg) Melting Pt: 149…153 °C Flash Pt: 293.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00235898 CAS Number: 86069-86-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...