Fmoc-L-homoleucine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Homoleu-OH, N-Fmoc-L-homoleucine, Fmoc-HoLeu-OH

A126536-250MG A126536-1G A126536-5G A126536-10G
103800-828EA 16 USD
103800-828 76797-680 77298-130 77298-132 77666-802
Fmoc-L-homoleucine 97%
Fmoc-L-homoleucine

Precaución: for Research Use Only

Formula: C₂₂H₂₅NO₄
Boiling Pt: 568.1 °C (760 mm Hg)
Flash Pt: 297.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00270206
CAS Number: 180414-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR