Fmoc-S-trityl-L-homocysteine 95% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: Fmoc-Homocys(Trt)-OH, Fmoc-HoCys(Trt)-OH

A147885-100MG A147885-250MG
103803-844EA 80 USD
103803-844 103841-958 77095-730 77095-732
Fmoc-S-trityl-L-homocysteine 95% (by HPLC)
Fmoc-S-trityl-L-homocysteine

Precaución: for Research Use Only

Formula: C₃₈H₃₃NO₄S
Boiling Pt: 780.1±60.0 °C (760 mm Hg)
Flash Pt: 425.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00672322
CAS Number: 167015-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR