Fmoc-HomoArg(Pbf)-OH 97% (by HPLC)

Supplier: Ambeed
Warning

A217879-1G A217879-100MG A217879-250MG A217879-10G A217879-100G
103818-664EA 27 USD
103818-664 103849-392 77088-034 77288-128 77288-130 77550-406 77615-834
Fmoc-HomoArg(Pbf)-OH 97% (by HPLC)
Fmoc-HomoArg(Pbf)-OH

Precaución: for Research Use Only

Formula: C₃₅H₄₂N₄O₇S
Storage Temperature: Refrigerator
MDL Number: MFCD03093571
CAS Number: 1159680-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR