N-Fmoc-glycine 98%

Supplier: Ambeed
Warning

Synonyms: 2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)acetic acid, Fmoc-Gly-OH

A423465-25G A423465-100G A423465-10G A423465-1G A423465-500G A423465-5G
103838-482EA 5 USD
103838-482 76842-118 77278-994 77278-996 77278-998 77279-000 77307-220
N-Fmoc-glycine 98%
N-Fmoc-glycine

Precaución: for Research Use Only

Formula: C₁₇H₁₅NO₄
MW: 297.3 g/mol
Boiling Pt: 545.7 °C (760 mm Hg)
Melting Pt: 174…178 °C
Flash Pt: 283.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00037140
CAS Number: 29022-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR