(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Glu-OH, N-Fmoc-L-glutamic Acid

A221584-100G A221584-10G A221584-500G A221584-5G A221584-25G
103818-936EA 6 USD
103818-936 76691-830 76691-832 76691-834 76691-836 77088-414
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid 95%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid

Precaución: for Research Use Only

Formula: C₂₀H₁₉NO₆
Boiling Pt: 594.3 °C (760 mm Hg)
Flash Pt: 338.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00237657
CAS Number: 121343-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR