Fmoc-Glu-OBzl 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Glu-Obzl, N-Fmoc-L-glutamic acid 1-benzyl ester

A119511-100G A119511-25G A119511-1G
103799-780EA 11 USD
103799-780 76971-422 76971-424 77311-898
Fmoc-Glu-OBzl 97%
Fmoc-Glu-OBzl

Precaución: for Research Use Only

Formula: C₂₇H₂₅NO₆
Boiling Pt: 702.9 °C (760 mm Hg)
Melting Pt: 117…121 °C
Flash Pt: 378.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00080272
CAS Number: 122350-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR