5-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate hydrate 98%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-L-glutamic acid 5-tert-butyl ester, Fmoc-Glu(OtBu)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamic Acid 5-tert-Butyl Ester, 5-tert-Butyl N-Fmoc-L-glutamate

A165615-100G A165615-10G A165615-500G A165615-25G A165615-5G
103808-958EA 597 USD
103808-958 103844-232 77273-520 77273-522 77273-524 77320-448 77320-450
5-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate hydrate 98%
5-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate hydrate

Precaución: for Research Use Only

Formula: C₂₄H₂₇NO₆
Melting Pt: 85…90 °C
Flash Pt: 339.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00037135
CAS Number: 71989-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR