Fmoc-Glu(OBzl)-OH 97%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-L-glutamic acid 5-benzyl ester

A138373-10G A138373-1G A138373-5G A138373-100G A138373-500G
103802-610EA 9 USD
103802-610 77272-356 77315-632 77315-634 77747-978 77747-979
Fmoc-Glu(OBzl)-OH 97%
Fmoc-Glu(OBzl)-OH

Precaución: for Research Use Only

Formula: C₂₇H₂₅NO₆
MW: 459.498 g/mol
Boiling Pt: 698.2 °C (760 mm Hg)
Melting Pt: 80…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065642
CAS Number: 123639-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR