Fmoc-Glu(OAll)-OH 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Fmoc-L-glutamic acid 5-allyl ester, N-Fmoc-L-glutamic acid 5-Allyl ester, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamic acid 5-Allyl ester, Fmoc-Glu(allyl ester)-OH, 5-Allyl N-Fmoc-L-glutamate, Fmoc-Glu(OAll)-OH, 5-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamate
A115243-100G
A115243-10G
A115243-250MG
A115243-25G
A115243-5G
A115243-1G
A115243-500G
103798-532EA
8
USD
103798-532
103824-382
76995-462
76995-464
76995-466
76995-468
77297-352
77311-196
77566-058
Fmoc-Glu(OAll)-OH 98%
Fmoc-Glu(OAll)-OH
Precaución: For research use only.
Formula:
C₂₃H₂₃NO₆ Melting Pt: 122…128 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00190879 CAS Number: 133464-46-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...