Fmoc-Glu(Edans)-OH 98%

Supplier: Ambeed
Warning

A157546-100MG A157546-250MG A157546-1G A157546-5G
103805-016EA 616 USD
103805-016 103843-216 76798-852 76798-854 77319-046 77319-048
Fmoc-Glu(Edans)-OH 98%
Fmoc-Glu(Edans)-OH

Precaución: for Research Use Only

Formula: C₃₂H₃₁N₃O₈S
Storage Temperature: Refrigerator
MDL Number: MFCD01076138
CAS Number: 193475-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR