Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamine 97%

Supplier: Ambeed
Warning

Synonyms: Nα-Fmoc-L-glutamine, Nɑ-Fmoc-L-glutamine, Fmoc-Gln-OH

A147878-100G A147878-10G A147878-500G A147878-25G A147878-5G
103841-956EA 6 USD
103841-956 77272-852 77272-854 77272-856 77299-608 77299-610
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamine 97%
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamine

Precaución: for Research Use Only

Formula: C₂₀H₂₀N₂O₅
MW: 368.39 g/mol
Melting Pt: 220 °C
Flash Pt: 377.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00037137
CAS Number: 71989-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR