Nα-Fmoc-N-δ-trityl-L-glutamine 98% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: Fmoc-Gln(Trt)-OH

A213511-5G A213511-100G A213511-10G A213511-500G A213511-1G A213511-25G
103818-302EA 7 USD
103818-302 103849-010 77087-526 77275-244 77275-246 77275-248 77303-290 77303-292
Nα-Fmoc-N-δ-trityl-L-glutamine 98% (by HPLC)
N-alpha-Fmoc-N-delta-trityl-L-glutamine

Precaución: for Research Use Only

Formula: C₃₉H₃₄N₂O₅
Melting Pt: 165…172 °C
Flash Pt: 482.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077056
CAS Number: 132327-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR