Fmoc-GABA-OH 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(Fmoc-amino)butyric acid, N-Fmoc-4-aminobutyric acid, Fmoc-γ-Abu-OH, Fmoc-GABA-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-aminobutyric acid
A174517-100G
A174517-10G
A174517-500G
A174517-1G
A174517-25G
A174517-5G
103806-950EA
7
USD
103806-950
103845-250
77151-442
77151-444
77151-446
77301-340
77301-342
77301-344
Fmoc-GABA-OH 97%
Fmoc-GABA-OH
Precaución: for Research Use Only
Formula:
C₁₉H₁₉NO₄ Boiling Pt: 571.9 °C (760 mm Hg) Melting Pt: 165…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00144889 CAS Number: 116821-47-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...