N-Fmoc-D-valine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-D-Val-OH, D-Valine, N-FMOC protected
A116281-100G
A116281-10G
A116281-25G
A116281-500G
A116281-1G
A116281-5G
103798-706EA
5
USD
103798-706
103824-568
77271-204
77271-206
77271-208
77271-210
77297-446
77311-384
N-Fmoc-D-valine 98%
N-Fmoc-D-valine
Precaución: for Research Use Only
Formula:
C₂₀H₂₁NO₄ Melting Pt: 143…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00062953 CAS Number: 84624-17-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...