(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid 96%

Supplier: Ambeed
Warning

Synonyms: O-tert-Butyl-Nalpha-Fmoc-D-tyrosine, Nalpha-Fmoc-O-tert-butyl-D-tyrosine, Fmoc-D-Tyr(tBu)-OH, N-Fmoc-O-tert-butyl-D-tyrosine

A239263-100G A239263-10G A239263-250MG A239263-25G A239263-5G A239263-1G
103811-272EA 9 USD
103811-272 103851-070 77000-368 77000-370 77000-372 77090-210 77090-212 77304-148
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid 96%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₈H₂₉NO₅
Boiling Pt: 658.2 °C (760 mm Hg)
Flash Pt: 351.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065684
CAS Number: 118488-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR