N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-serine 97%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-D-serine, Fmoc-D-Ser-OH

A155152-100G A155152-10G A155152-25G A155152-1G A155152-5G
103804-724EA 5 USD
103804-724 103842-920 77028-342 77028-344 77028-346 77318-646 77318-648
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-serine 97%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-serine

Precaución: for Research Use Only

Formula: C₁₈H₁₇NO₅
MW: 327.34 g/mol
Boiling Pt: 599.3 °C (760 mm Hg)
Melting Pt: 108…112 °C
Flash Pt: 316.248 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077068
CAS Number: 116861-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR