Fmoc-O-tert-butyl-D-serine 97%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-O-tert-butyl-D-serine, Fmoc-D-Ser(tBu)-OH

A201780-100G A201780-10G A201780-250MG A201780-500G A201780-25G A201780-1G
103817-330EA 9 USD
103817-330 103847-970 76999-348 76999-350 76999-352 76999-354 77086-054 77302-780
Fmoc-O-tert-butyl-D-serine 97%
Fmoc-O-tert-butyl-D-serine

Precaución: for Research Use Only

Formula: C₂₂H₂₅NO₅
MW: 383.4 g/mol
Boiling Pt: 578.6 °C (760 mm Hg)
Flash Pt: 303.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077071
CAS Number: 128107-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR