Fmoc-D-Ser(Bzl)-OH 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Fmoc-O-benzyl-D-serine
A320950-100G
A320950-5G
A320950-1G
103825-260EA
7
USD
103825-260
103836-192
76802-902
76839-080
77306-232
Fmoc-D-Ser(Bzl)-OH 97%
Fmoc-D-Ser(Bzl)-OH
Precaución: for Research Use Only
Formula:
C₂₅H₂₃NO₅ Melting Pt: 150…154 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00237032 CAS Number: 122889-11-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...