Fmoc-D-Phg-OH 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-D-Phg-OH, N-Fmoc-D-α-phenylglycine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-2-phenylglycine, N-Fmoc-D-2-phenylglycine
A101674-10G
A101674-25G
A101674-1G
A101674-100G
103796-082EA
8
USD
103796-082
103821-778
77148-228
77148-230
77308-700
77610-750
Fmoc-D-Phg-OH 98%
Fmoc-D-Phg-OH
Precaución: for Research Use Only
Formula:
C₂₃H₁₉NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00190891 CAS Number: 111524-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...