Fmoc-D-phenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Phe-OH, N-Fmoc-D-phenylalanine

A148507-100G A148507-10G A148507-500G A148507-25G
103842-050EA 7 USD
103842-050 77027-994 77027-996 77027-998 77317-510
Fmoc-D-phenylalanine 98%
Fmoc-D-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₂₁NO₄
MW: 387.4 g/mol
Boiling Pt: 620.1 °C (760 mm Hg)
Melting Pt: 181…185 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00062955
CAS Number: 86123-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR