N-Fmoc-4-iodo-D-phenylalanine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-D-Phe(4-I)-OH
A133945-250MG
A133945-1G
A133945-5G
A133945-10G
103801-938EA
15
USD
103801-938
76996-546
77314-638
77314-640
77670-914
N-Fmoc-4-iodo-D-phenylalanine 97%
N-Fmoc-4-iodo-D-phenylalanine
Precaución: for Research Use Only
Formula:
C₂₄H₂₀INO₄ MW: 513.331 g/mol Boiling Pt: 658.8 °C (760 mm Hg) Flash Pt: 352.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00672557 CAS Number: 205526-29-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...