Fmoc-3,4-dichloro-D-phenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Phe(3,4-DiCl)-OH, Fmoc-3,4-dichloro-D-Phe-OH

A100391-250MG A100391-25G A100391-5G A100391-1G
103795-414EA 23 USD
103795-414 77025-130 77025-132 77025-134 77282-488
Fmoc-3,4-dichloro-D-phenylalanine 95%
Fmoc-3,4-dichloro-D-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₁₉Cl₂NO₄
MW: 456.319 g/mol
Boiling Pt: 661.7 °C (760 mm Hg)
Melting Pt: 122 °C
Flash Pt: 354 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00273472
CAS Number: 177966-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR