Fmoc-D-Orn(Boc)-OH 96%

Supplier: Ambeed
Warning

Synonyms: Nδ-Boc-Nα-Fmoc-D-ornithine

A359427-10G A359427-1G A359427-5G
103826-228EA 40 USD
103826-228 103837-236 76803-448 76840-408 76840-410
Fmoc-D-Orn(Boc)-OH 96%
Fmoc-D-Orn(Boc)-OH

Precaución: for Research Use Only

Formula: C₂₅H₃₀N₂O₆
MW: 454.523 g/mol
Boiling Pt: 679 °C (760 mm Hg)
Flash Pt: 364.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077065
CAS Number: 118476-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR