N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-methionine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Fmoc-D-methionine, Fmoc-D-Met-OH
A291379-25G
A291379-5G
A291379-100G
A291379-10G
A291379-1G
103813-914EA
9
USD
103813-914
103835-054
76837-648
76837-650
77277-170
77277-172
77305-612
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-methionine 97%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-methionine
Precaución: for Research Use Only
Formula:
C₂₀H₂₁NO₄S MW: 371.45 g/mol Melting Pt: 120…124 °C Flash Pt: 325.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00062958 CAS Number: 112883-40-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...