Fmoc-D-Lys(Alloc)-OH 97%

Supplier: Ambeed
Warning

Synonyms: Nepsilon-Allyloxycarbonyl-Nα-Fmoc-D-lysine, Fmoc-D-Lys(Aloc)-OH

A276328-250MG A276328-1G A276328-5G A276328-100G A276328-10G
103813-302EA 16 USD
103813-302 103853-234 77002-074 77093-086 77093-088 77672-418 77672-420
Fmoc-D-Lys(Alloc)-OH 97%
Fmoc-D-Lys(Alloc)-OH

Precaución: for Research Use Only

Formula: C₂₅H₂₈N₂O₆
MW: 452.507 g/mol
Boiling Pt: 689.7 °C (760 mm Hg)
Flash Pt: 370.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00798639
CAS Number: 214750-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR