N-Fmoc-D-leucine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Leu-OH

A142660-10G A142660-1G A142660-5G A142660-100G A142660-500G
103803-202EA 5 USD
103803-202 77272-540 77316-440 77316-442 77549-724 77748-000
N-Fmoc-D-leucine 97%
N-Fmoc-D-leucine

Precaución: for Research Use Only

Formula: C₂₁H₂₃NO₄
MW: 353.4 g/mol
Boiling Pt: 559.8 °C (760 mm Hg)
Melting Pt: 155 °C
Flash Pt: 292.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00062957
CAS Number: 114360-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR