Fmoc-d-pipecolic acid 97%

Supplier: Ambeed
Warning

Synonyms: (R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-2-carboxylic acid, 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-pipecolic acid, (R)-1-Fmoc-2-piperidinecarboxylic acid, Fmoc-D-Pip-OH, 1-Fmoc-D-pipecolic acid, Fmoc-D-Homopro-OH, (R)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid

A255696-250MG A255696-1G A255696-5G A255696-10G
103812-206EA 14 USD
103812-206 103852-066 76801-678 77091-524 77091-526 77879-471
Fmoc-d-pipecolic acid 97%
Fmoc-d-pipecolic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₄
Boiling Pt: 561.6 °C (760 mm Hg)
Melting Pt: 154…158 °C
Flash Pt: 293.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00235899
CAS Number: 101555-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR