Fmoc-D-Homophe-OH 97%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-D-homophenylalanine, Fmoc-D-Homophe-OH, Fmoc-D-HoPhe-OH

A204134-250MG A204134-1G A204134-25G A204134-5G
103817-528EA 12 USD
103817-528 103848-192 76999-440 77086-344 77086-346 77086-348
Fmoc-D-Homophe-OH 97%
Fmoc-D-Homophe-OH

Precaución: for Research Use Only

Formula: C₂₅H₂₃NO₄
Boiling Pt: 628.3 °C (760 mm Hg)
Flash Pt: 333.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151917
CAS Number: 135944-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR