Fmoc-D-Glu-OtBu 95%

Supplier: Ambeed
Warning

A289318-100G A289318-250MG A289318-1G A289318-25G A289318-5G A289318-10G
103813-824EA 16 USD
103813-824 103834-964 77002-328 77002-330 77305-556 77305-558 77305-560 77881-799
Fmoc-D-Glu-OtBu 95%
Fmoc-D-Glu-OtBu

Precaución: for Research Use Only

Formula: C₂₄H₂₇NO₆
Storage Temperature: Refrigerator
MDL Number: MFCD00077054
CAS Number: 109745-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR