N-Fmoc-N'-trityl-D-glutamine 97%

Supplier: Ambeed
Warning

Synonyms: Nα-Fmoc-Nδ-trityl-D-glutamine, Fmoc-D-Gln(Trt)-OH

A152894-100G A152894-10G A152894-250MG A152894-1G A152894-5G
103804-446EA 19 USD
103804-446 103842-618 76997-422 76997-424 76997-426 77318-250 77318-252
N-Fmoc-N'-trityl-D-glutamine 97%
N-Fmoc-N'-trityl-D-glutamine

Precaución: for Research Use Only

Formula: C₃₉H₃₄N₂O₅
MW: 610.71 g/mol
Boiling Pt: 873.5 °C (760 mm Hg)
Melting Pt: 232…237 °C
Flash Pt: 482.1 °C
Storage Temperature: Freezer
MDL Number: MFCD00151924
CAS Number: 200623-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR