Fmoc-D-Dap(Boc)-OH 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid
A111315-250MG
A111315-1G
A111315-100G
A111315-10G
103797-816EA
11
USD
103797-816
103823-632
77094-010
77310-436
77900-874
77901-877
Fmoc-D-Dap(Boc)-OH 95%
Fmoc-D-Dap(Boc)-OH
Precaución: for Research Use Only
Formula:
C₂₃H₂₆N₂O₆ MW: 426.469 g/mol Boiling Pt: 652.5±55.0 °C (760 mm Hg) Flash Pt: 348.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00798648 CAS Number: 198544-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...