Fmoc-D-Dab-OH 97%

Supplier: Ambeed
Warning

Synonyms: (R)-2-(Fmoc-amino)-4-aminobutyric acid

A101955-250MG A101955-1G A101955-5G
103796-224EA 90 USD
103796-224 103821-918 77093-536 77308-804 77308-806
Fmoc-D-Dab-OH 97%
Fmoc-D-Dab-OH

Precaución: for Research Use Only

Formula: C₁₉H₂₀N₂O₄
MW: 340.379 g/mol
Boiling Pt: 593.7 °C (760 mm Hg)
Flash Pt: 312.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00237018
CAS Number: 201484-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR